Assay Details
Target Gene Details
Entrez Gene ID: | 99011 |
Gene Name: | protein-O-mannosyltransferase 1 |
Gene Aliases: |
AI505244 |
Location: |
Chr.2:32236595-32255005 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pomt1 | NM_145145.1 | NP_660127.1 | ||
| AK053889.1 | BAC35577.1 | |||
| AK134770.1 | BAE22274.1 | |||
| AK153984.1 | BAE32295.1 | |||
| AY007238.1 | AAG15588.1 | |||
| AY494857.1 | AAS76201.1 | |||
| BC027325.1 | AAH27325.1 |
More Information
Set Membership: |
|

Genomic Map