Assay Details
Target Gene Details
Entrez Gene ID: | 18779 |
Gene Name: | phospholipase A2 receptor 1 |
Gene Aliases: |
Pla2-r, Pla2g1br, Pla2r |
Location: |
Chr.2:60417543-60553350 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2r1 | NM_008867.2 | NP_032893.1 | ||
| BC141355.1 | ||||
| BC141356.1 | ||||
| D30779.1 | BAA06443.1 |

Genomic Map