Assay Details
Target Gene Details
Entrez Gene ID: | 65112 |
Gene Name: | prostate transmembrane protein, androgen induced 1 |
Gene Aliases: |
2210418I02Rik, AW455466, Erg1.2, N4wbp4, Stag1, Tmepai |
Location: |
Chr.2:173224458-173276533 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmepa1 | NM_022995.3 | NP_075371.2 | ||
| AF220208.1 | AAG44247.1 | |||
| AK008976.1 | BAB26001.1 | |||
| AK136450.1 | ||||
| BC036995.1 | AAH36995.1 |

Genomic Map