Assay Details
Target Gene Details
Entrez Gene ID: | 192166 |
Gene Name: | sarcosine dehydrogenase |
Gene Aliases: |
- |
Location: |
Chr.2:27188393-27248332 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sardh | NM_138665.2 | 3 | 1094 | NP_619606.1 |
| AK140451.1 | 2 | 370 | ||
| AK159236.1 | 3 | 1058 | BAE34920.1 | |
| AK159681.1 | 3 | 1058 | BAE35284.1 | |
| AK159704.1 | 3 | 1058 | BAE35303.1 | |
| AK163515.1 | 3 | 623 | BAE37376.1 | |
| AK163660.1 | 2 | 370 | BAE37445.1 | |
| AK213849.1 | ||||
| BC003456.1 | 3 | 603 | AAH03456.1 |
More Information
Set Membership: |
|

Genomic Map