Assay Details
Target Gene Details
Entrez Gene ID: | 30963 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 1 |
Gene Aliases: |
Ptpla |
Location: |
Chr.2:14026366-14056035 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hacd1 | NM_001012396.2 | NP_001012396.2 | ||
| NM_013935.3 | NP_038963.3 | |||
| AK004461.1 | BAE43179.1 | |||
| AK010119.1 | BAB26713.1 | |||
| AK076818.1 | ||||
| BB662124.1 | ||||
| BC138116.1 | ||||
| BC141017.1 | ||||
| BE633820.1 | ||||
| CF751799.1 |

Genomic Map