Assay Details
Target Gene Details
Entrez Gene ID: | 66461 |
Gene Name: | protein tyrosine phosphatase, mitochondrial 1 |
Gene Aliases: |
1110001D10Rik, 2810004N20Rik, PLIP |
Location: |
Chr.2:90910713-90918050 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptpmt1 | NM_025576.2 | NP_079852.1 | ||
AK003210.1 | ||||
AK012674.1 | BAB28400.1 | |||
AK014691.1 | BAB29504.1 | |||
AK151972.1 | BAE30841.1 | |||
BC026750.1 | AAH26750.1 |
