Assay Details
Target Gene Details
Entrez Gene ID: | 319817 |
Gene Name: | ring finger and CCCH-type zinc finger domains 2 |
Gene Aliases: |
2900024N03Rik, 9430019J22Rik, D930043C02Rik, Mnab, Rnf164 |
Location: |
Chr.2:37368861-37423380 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rc3h2 | NM_001100591.1 | 14 | 2644 | NP_001094061.1 |
| NM_001290642.1 | 14 | 2644 | NP_001277571.1 | |
| AK053071.1 | 14 | 2644 | ||
| BC117951.1 | 14 | 2637 | ||
| BC117952.1 | 14 | 2637 |

Genomic Map