Assay Details
Target Gene Details
Entrez Gene ID: | 68475 |
Gene Name: | Sjogren's syndrome nuclear autoantigen 1 |
Gene Aliases: |
1110003H09Rik, 1190004J23Rik, AA934184, N14, NA14 |
Location: |
Chr.2:25271039-25272418 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssna1 | NM_023464.2 | NP_075953.1 | ||
| AB041656.1 | BAA95103.1 | |||
| AK003356.2 | BAB22733.2 | |||
| AK088418.1 | BAC40344.1 | |||
| BC037081.1 | AAH37081.1 | |||
| BC048360.1 | AAH48360.1 | |||
| BY703613.1 |

Genomic Map