Assay Details
Target Gene Details
Entrez Gene ID: | 269400 |
Gene Name: | regulator of telomere elongation helicase 1 |
Gene Aliases: |
AI451565, AW540478, Rtel |
Location: |
Chr.2:181288028-181356616 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtel1 | NM_001001882.3 | NP_001001882.3 | ||
| NM_001166665.1 | NP_001160137.1 | |||
| NM_001166666.1 | NP_001160138.1 | |||
| NM_001166667.1 | NP_001160139.1 | |||
| NM_001166668.1 | NP_001160140.1 | |||
| NR_030710.1 | ||||
| AK142225.1 | ||||
| AK145145.1 | BAE26258.1 | |||
| AK173097.1 | BAD32375.1 | |||
| AY481619.1 | AAR27234.1 | |||
| AY481620.1 | AAR27235.1 | |||
| AY481621.1 | AAR27236.1 | |||
| AY481622.1 | AAR27237.1 | |||
| AY481623.1 | AAR27238.1 | |||
| BC105578.1 | ||||
| BC144977.1 | ||||
| BC144978.1 | ||||
| BC145658.1 |
More Information
Set Membership: |
|

Genomic Map