Assay Details
Target Gene Details
Entrez Gene ID: | 241770 |
Gene Name: | regulating synaptic membrane exocytosis 4 |
Gene Aliases: |
6530406L02, Rim4 |
Location: |
Chr.2:163859756-163918990 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rims4 | NM_183023.1 | NP_898844.1 | ||
| AK134613.1 | BAE22208.1 | |||
| AY326955.1 | AAQ01682.1 | |||
| BC115475.1 |
More Information
Set Membership: |
|

Genomic Map