Assay Details
Target Gene Details
Entrez Gene ID: | 20910 |
Gene Name: | syntaxin binding protein 1 |
Gene Aliases: |
AI317162, AI326233, MMS10-G, Ms10g, Munc-18a, Munc18-1, N-sec1, Rb-sec1, Sxtbp1, Unc18-1, Unc18h, nsec1 |
Location: |
Chr.2:32787607-32847237 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stxbp1 | NM_001113569.1 | NP_001107041.1 | ||
| NM_009295.2 | NP_033321.2 | |||
| AF326545.1 | AAL37391.1 | |||
| AF326563.1 | AAL37409.1 | |||
| BB852672.1 | ||||
| BC031728.1 | AAH31728.1 | |||
| D45903.1 | BAA19479.1 |
More Information
Set Membership: |
|

Genomic Map