Assay Details
Target Gene Details
Entrez Gene ID: | 74182 |
Gene Name: | glycerophosphocholine phosphodiesterase GDE1 homolog (S. cerevisiae) |
Gene Aliases: |
2310004G06Rik, 2310032D16Rik, AU015213, Gde5, Prei4, mKIAA1434 |
Location: |
Chr.2:132529082-132578282 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpcpd1 | NM_001042672.1 | NP_001036137.1 | ||
| NM_001291050.1 | NP_001277979.1 | |||
| NM_001291051.1 | NP_001277980.1 | |||
| NM_001291052.1 | NP_001277981.1 | |||
| NM_027096.2 | NP_081372.1 | |||
| NM_028802.3 | NP_083078.3 | |||
| AK030645.1 | BAC27063.1 | |||
| AK031658.1 | ||||
| AK050318.1 | ||||
| AK141374.1 | ||||
| AK163419.1 | ||||
| AK165952.1 | ||||
| AK166454.1 | ||||
| BC033408.1 | AAH33408.1 |
More Information
Set Membership: |
|

Genomic Map