Assay Details
Target Gene Details
Entrez Gene ID: | 20687 |
Gene Name: | trans-acting transcription factor 3 |
Gene Aliases: |
D130027J01Rik |
Location: |
Chr.2:72936430-72980446 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sp3 | NM_001018042.3 | NP_001018052.1 | ||
| BC079874.1 | AAH79874.1 |
Target Gene Details
Entrez Gene ID: | 100503849 |
Gene Name: | trans-acting transcription factor 3, opposite strand |
Gene Aliases: |
1700011J10Rik |
Location: |
Chr.2:72979432-72989249 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sp3os | NR_045269.2 | |||
| AK005870.1 | BAC25127.1 | |||
| AK013593.1 | ||||
| BY152393.1 |
More Information
Set Membership: |
|

Genomic Map