Assay Details
Target Gene Details
Entrez Gene ID: | 228765 |
Gene Name: | syndecan binding protein (syntenin) 2 |
Gene Aliases: |
- |
Location: |
Chr.2:151572609-151590005 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdcbp2 | NM_145535.2 | NP_663510.1 | ||
| AK136572.1 | BAE23054.1 | |||
| AK144452.1 | BAE25896.1 | |||
| AK160483.1 | BAE35814.1 | |||
| BC005556.1 | AAH05556.1 |
More Information
Set Membership: |
|

Genomic Map