Assay Details
Target Gene Details
Entrez Gene ID: | 228765 |
Gene Name: | syndecan binding protein (syntenin) 2 |
Gene Aliases: |
- |
Location: |
Chr.2:151572609-151590005 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdcbp2 | NM_145535.2 | 7 | 743 | NP_663510.1 |
| AK136572.1 | 7 | 699 | BAE23054.1 | |
| AK144452.1 | 7 | 708 | BAE25896.1 | |
| AK160483.1 | 7 | 701 | BAE35814.1 | |
| BC005556.1 | 7 | 734 | AAH05556.1 |
More Information
Set Membership: |
|

Genomic Map