Assay Details
Target Gene Details
Entrez Gene ID: | 19182 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, ATPase 3 |
Gene Aliases: |
TBP-1 |
Location: |
Chr.2:91054016-91059438 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmc3 | NM_008948.2 | NP_032974.2 | ||
| AK002542.1 | ||||
| AK151370.1 | BAE30344.1 | |||
| AK152018.1 | BAE30880.1 | |||
| AK167002.1 | BAE39181.1 | |||
| AK167061.1 | BAE39223.1 | |||
| AK168263.1 | BAE40211.1 | |||
| AK168616.1 | BAE40481.1 | |||
| BC005783.1 | AAH05783.1 | |||
| BE989849.1 | ||||
| D49686.1 | BAA32559.1 |
More Information
Set Membership: |
|

Genomic Map