Assay Details
Target Gene Details
Entrez Gene ID: | 19074 |
Gene Name: | proteoglycan 2, bone marrow |
Gene Aliases: |
MBP, mMBP, mMBP-1 |
Location: |
Chr.2:84980461-84983632 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prg2 | NM_008920.4 | NP_032946.1 | ||
| AK010901.1 | BAB27254.1 | |||
| AK156715.1 | BAE33816.1 | |||
| BC109348.2 | AAI09349.1 |
More Information
Set Membership: |
|

Genomic Map