Assay Details
Target Gene Details
Entrez Gene ID: | 68365 |
Gene Name: | RAB14, member RAS oncogene family |
Gene Aliases: |
0610030G24Rik, 2810475J17Rik, A830021G03Rik, AI314285, AI649155, D030017L14Rik |
Location: |
Chr.2:35180205-35201431 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab14 | NM_026697.3 | NP_080973.1 | ||
| AB232609.1 | BAF02871.1 | |||
| AJ968955.1 | CAI92125.1 | |||
| AJ968956.1 | CAI92126.1 | |||
| AJ968957.1 | CAI92127.1 | |||
| AJ968958.1 | CAI92128.1 | |||
| AJ968959.1 | CAI92129.1 | |||
| AJ968960.1 | CAI92130.1 | |||
| AK002704.1 | BAB22298.1 | |||
| AK013410.1 | ||||
| AK133963.1 | BAE21957.1 | |||
| AK135751.1 | ||||
| AK160859.1 | BAE36051.1 | |||
| AK165812.1 | BAE38391.1 | |||
| AK168551.1 | BAE40426.1 | |||
| AK168867.1 | BAE40686.1 | |||
| AK169396.1 | BAE41143.1 | |||
| AK181684.1 | ||||
| AK186545.1 | ||||
| AK216803.1 | ||||
| BC009085.1 | AAH09085.1 | |||
| BC025139.1 | AAH25139.1 | |||
| BC056648.1 | AAH56648.1 |

Genomic Map