Assay Details
Target Gene Details
Entrez Gene ID: | 20779 |
Gene Name: | Rous sarcoma oncogene |
Gene Aliases: |
AW259666, pp60c-src |
Location: |
Chr.2:157423901-157471838 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Src | NM_001025395.2 | 8 | 965 | NP_001020566.1 |
| NM_009271.3 | 9 | 983 | NP_033297.2 | |
| AK017629.1 | 1 | 1111 | ||
| AK201096.1 | ||||
| BC039953.1 | 8 | 975 | AAH39953.1 | |
| CX218263.1 | 5 | 464 | ||
| M17031.1 | 6 | 599 | AAA40135.1 |

Genomic Map