Assay Details
Target Gene Details
Entrez Gene ID: | 56878 |
Gene Name: | RNA binding motif, single stranded interacting protein 1 |
Gene Aliases: |
2600014B10Rik, AI255215, MSSP-1, MSSP-2, MSSP-3, YC1 |
Location: |
Chr.2:60750193-60963450 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbms1 | NM_001141931.1 | 11 | 1247 | NP_001135403.1 |
| NM_001141932.1 | 12 | 1295 | NP_001135404.1 | |
| NM_020296.2 | 11 | 1440 | NP_064692.2 | |
| AB026569.1 | 11 | 1163 | BAA77262.1 | |
| AK133054.1 | 11 | 968 | BAE21490.1 | |
| AK135280.1 | 11 | 1127 | BAE22471.1 | |
| AK145897.1 | 11 | 1249 | BAE26733.1 | |
| AK147109.1 | 11 | 1441 | BAE27681.1 | |
| AK161102.1 | 12 | 1245 | BAE36195.1 | |
| AK161416.1 | 7 | 577 | BAE36384.1 | |
| AK178547.1 | 6 | 327 | ||
| AK180025.1 | 4 | 279 | ||
| AK188295.1 | 5 | 278 | ||
| AK203574.1 | 3 | 194 | ||
| AK210454.1 | 6 | 356 | ||
| AK218973.1 | 3 | 186 | ||
| BC016501.1 | 11 | 1428 | ||
| BC056609.1 | 9 | 687 | AAH56609.1 | |
| BC057866.1 | 12 | 1327 | AAH57866.1 |

Genomic Map