Assay Details
Target Gene Details
Entrez Gene ID: | 20459 |
Gene Name: | PTK6 protein tyrosine kinase 6 |
Gene Aliases: |
BRK, Sik, Tksk, tks |
Location: |
Chr.2:181195124-181203810 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptk6 | NM_009184.2 | 7 | 1375 | NP_033210.1 |
| BC125342.1 | 7 | 1346 | ||
| BC125344.1 | 7 | 1346 | ||
| S74391.1 | 2 | 169 | AAB32402.2 | |
| U16805.1 | 7 | 1375 | AAA67929.1 |

Genomic Map