Assay Details
Target Gene Details
Entrez Gene ID: | 76927 |
Gene Name: | TSSK6 activating co-chaperone |
Gene Aliases: |
1700021C14Rik, SIP |
Location: |
Chr.3:88282752-88297009 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsacc | NM_029801.2 | 4 | 335 | NP_084077.1 |
| AK006195.1 | 4 | 336 | BAB24452.1 | |
| BC107298.2 | 4 | 301 | AAI07299.1 |
More Information
Set Membership: |
|

Genomic Map