Assay Details
Target Gene Details
Entrez Gene ID: | 74410 |
Gene Name: | tubulin tyrosine ligase-like family, member 11 |
Gene Aliases: |
4932702F08Rik, 4933424A20Rik, AU043211, D2Ertd624e |
Location: |
Chr.2:35751226-35979624 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll11 | NM_029774.2 | 8 | 1777 | NP_084050.1 |
| AK016577.1 | 7 | 1135 | BAB30318.1 | |
| AM690755.1 | 8 | 1777 | ||
| BC115858.1 | 5 | 1127 |
More Information
Set Membership: |
|

Genomic Map