Assay Details
Target Gene Details
Entrez Gene ID: | 75740 |
Gene Name: | EGF-like and EMI domain containing 1 |
Gene Aliases: |
6130401L20Rik, Efcab11 |
Location: |
Chr.3:29080745-29691058 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Egfem1 | NM_001167748.1 | NP_001161220.1 | ||
| NM_029412.1 | NP_083688.1 | |||
| AK018073.1 | BAB31061.1 | |||
| AK032017.1 | BAC27650.1 | |||
| BC117810.1 |
More Information
Set Membership: |
|

Genomic Map