Assay Details
Target Gene Details
Entrez Gene ID: | 54401 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide |
Gene Aliases: |
1300003C17Rik |
Location: |
Chr.2:163995197-164018588 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ywhab | NM_018753.6 | NP_061223.2 | ||
| AF058797.1 | AAC14343.1 | |||
| AK002632.1 | BAB22246.1 | |||
| AK004872.1 | BAB23631.1 | |||
| AK011389.1 | BAB27587.1 | |||
| AK083367.1 | BAC38886.1 | |||
| AK144061.1 | BAE25678.1 | |||
| AK150414.1 | BAE29538.1 | |||
| AK151294.1 | BAE30278.1 | |||
| AK158932.1 | BAE34730.1 | |||
| AK178774.1 | ||||
| AK186637.1 | ||||
| AK188420.1 | ||||
| AK206680.1 | ||||
| AK206731.1 | ||||
| AK213523.1 | ||||
| AK217062.1 | ||||
| AK217335.1 | ||||
| AK219160.1 | ||||
| BC132121.1 | ||||
| BC138512.1 |
More Information
Set Membership: |
|

Genomic Map