Assay Details
Target Gene Details
Entrez Gene ID: | 228012 |
Gene Name: | tousled-like kinase 1 |
Gene Aliases: |
4930545J15Rik |
Location: |
Chr.2:70712407-70825480 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tlk1 | NM_172664.3 | NP_766252.2 | ||
| AK029773.1 | BAC26610.1 | |||
| AK045308.1 | ||||
| BC033931.1 | ||||
| BC051641.1 | AAH51641.1 |
More Information
Set Membership: |
|

Genomic Map