Assay Details
Target Gene Details
Entrez Gene ID: | 114889 |
Gene Name: | visual system homeobox 1 homolog (zebrafish) |
Gene Aliases: |
CHX10-like |
Location: |
Chr.2:150680702-150689137 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vsx1 | NM_054068.2 | NP_473409.1 | ||
| AF391758.1 | AAL09585.1 | |||
| AF395732.1 | AAL08947.1 | |||
| AK044393.1 | ||||
| AK044687.1 | ||||
| AK157311.1 | BAE34043.1 | |||
| AY056825.1 | AAL11431.1 | |||
| BC057647.1 | AAH57647.1 | |||
| BC138456.1 | ||||
| BC138460.1 |
More Information
Set Membership: |
|

Genomic Map