Assay Details
Target Gene Details
Entrez Gene ID: | 639634 |
Gene Name: | arylacetamide deacetylase-like 2 |
Gene Aliases: |
EG639634 |
Location: |
Chr.3:60006743-60025420 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aadacl2 | NM_001128091.1 | 2 | 515 | NP_001121563.1 |
| BC147724.1 | 2 | 515 | ||
| BC147728.1 | 2 | 515 |
More Information
Set Membership: |
|

Genomic Map