Assay Details
Target Gene Details
Entrez Gene ID: | 22174 |
Gene Name: | TYRO3 protein tyrosine kinase 3 |
Gene Aliases: |
AI323366, Brt, Dtk, Etk-2, Rse, Sky, TK19-2, Tif, etk2/tyro3 |
Location: |
Chr.2:119797740-119818104 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tyro3 | NM_001290800.1 | 2 | 973 | NP_001277729.1 |
| NM_019392.2 | 2 | 509 | NP_062265.2 | |
| AB000828.1 | 2 | 246 | BAA19193.1 | |
| AK141198.1 | 2 | 280 | BAE24581.1 | |
| BC066058.1 | 2 | 509 | AAH66058.1 | |
| BC082325.1 | 2 | 457 | AAH82325.1 | |
| D17393.1 | 2 | 954 | BAA04216.1 | |
| U05683.1 | 2 | 294 | AAA19237.1 | |
| U18342.1 | 2 | 426 | AAB26942.1 | |
| U18343.1 | 2 | 969 | AAB26943.1 | |
| U18933.1 | 2 | 445 | AAC52148.1 | |
| X78103.1 | 2 | 453 | CAA54995.1 |
More Information
Set Membership: |
|

Genomic Map