Assay Details
Target Gene Details
Entrez Gene ID: | 22174 |
Gene Name: | TYRO3 protein tyrosine kinase 3 |
Gene Aliases: |
AI323366, Brt, Dtk, Etk-2, Rse, Sky, TK19-2, Tif, etk2/tyro3 |
Location: |
Chr.2:119797740-119818104 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tyro3 | NM_001290800.1 | NP_001277729.1 | ||
| NM_019392.2 | NP_062265.2 | |||
| AB000828.1 | BAA19193.1 | |||
| AK141198.1 | BAE24581.1 | |||
| BC066058.1 | AAH66058.1 | |||
| BC082325.1 | AAH82325.1 | |||
| D17393.1 | BAA04216.1 | |||
| U05683.1 | AAA19237.1 | |||
| U18342.1 | AAB26942.1 | |||
| U18343.1 | AAB26943.1 | |||
| U18933.1 | AAC52148.1 | |||
| X78103.1 | CAA54995.1 |
More Information
Set Membership: |
|

Genomic Map