Assay Details
Target Gene Details
Entrez Gene ID: | 228911 |
Gene Name: | teashirt zinc finger family member 2 |
Gene Aliases: |
2900073F20Rik, B830045G17, Mtsh2, Sdccag33l, Tsh2, Zfp218, mKIAA4248, teashirt2 |
Location: |
Chr.2:169632496-170071666 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tshz2 | NM_080455.2 | NP_536703.2 | ||
| BC079550.1 | AAH79550.1 | |||
| BC079877.1 | 1 | 368 | AAH79877.1 | |
| CN670346.1 |
More Information
Set Membership: |
|

Genomic Map