Assay Details
Target Gene Details
Entrez Gene ID: | 21400 |
Gene Name: | transcription elongation factor A (SII), 2 |
Gene Aliases: |
AI326274, S-II-T1, SII-T1, Tceat |
Location: |
Chr.2:181680281-181688069 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcea2 | NM_009326.2 | NP_033352.1 | ||
| AB010271.1 | BAA82313.1 | |||
| AB221557.1 | ||||
| AK131650.1 | ||||
| AK137527.1 | BAE23395.1 | |||
| BC049617.1 | ||||
| BC065786.1 | ||||
| BC138504.1 | ||||
| BC138505.1 | ||||
| D86081.1 | BAA19752.1 |
More Information
Set Membership: |
|

Genomic Map