Assay Details
Target Gene Details
Entrez Gene ID: | 96957 |
Gene Name: | transmembrane protein 62 |
Gene Aliases: |
B830009D23Rik, C77355 |
Location: |
Chr.2:120976719-121007849 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem62 | NM_175285.3 | 5 | 785 | NP_780494.1 |
| AK046793.1 | 5 | 584 | BAC32871.1 | |
| AK161890.1 | 5 | 835 | ||
| BC070410.1 | 5 | 785 | AAH70410.1 |
More Information
Set Membership: |
|

Genomic Map