Assay Details
Target Gene Details
Entrez Gene ID: | 629761 |
Gene Name: | WAP four-disulfide core domain 11 |
Gene Aliases: |
- |
Location: |
Chr.2:164662894-164674253 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wfdc11 | NM_001161806.1 | NP_001155278.1 | ||
| AK079019.1 |
More Information
Set Membership: |
|

Genomic Map