Assay Details
Target Gene Details
Entrez Gene ID: | 192200 |
Gene Name: | WAP four-disulfide core domain 12 |
Gene Aliases: |
Swam2, WAP2 |
Location: |
Chr.2:164189231-164190558 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wfdc12 | NM_138684.2 | NP_619625.1 | ||
| AF276975.1 | AAF86472.1 | |||
| AK008999.1 | BAE20421.1 | |||
| BC030939.1 | AAH30939.1 |
More Information
Set Membership: |
|

Genomic Map