Assay Details
Target Gene Details
Entrez Gene ID: | 140629 |
Gene Name: | U box domain containing 5 |
Gene Aliases: |
1500010O06Rik, C330018L13Rik, Rnf37, UIP5, Ubce7ip5, Ube7ip5 |
Location: |
Chr.2:130589996-130630038 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubox5 | NM_001255993.1 | 4 | 474 | NP_001242922.1 |
| NM_001255994.1 | 3 | 327 | NP_001242923.1 | |
| NM_080562.5 | 3 | 332 | NP_542129.2 | |
| AF360997.1 | 3 | 262 | AAK51467.1 | |
| AK005195.1 | 3 | 293 | ||
| AK034114.1 | 3 | 281 | BAC28590.1 | |
| AK049276.1 | 2 | 98 | BAC33652.1 | |
| AK158052.1 | 4 | 432 | BAE34338.1 | |
| AK170024.1 | 4 | 2175 | ||
| AK170100.1 | 3 | 303 | ||
| AK171070.1 | 4 | 441 | ||
| BC025068.1 | 2 | 194 | AAH25068.1 |

Genomic Map