Assay Details
Target Gene Details
Entrez Gene ID: | 66715 |
Gene Name: | HEN1 methyltransferase homolog 1 (Arabidopsis) |
Gene Aliases: |
4921515J06Rik, Hen1, mHEN1 |
Location: |
Chr.3:108938542-108960776 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Henmt1 | NM_001078646.1 | NP_001072114.1 | ||
| NM_025723.2 | NP_079999.2 | |||
| AK014900.1 | ||||
| AK038994.1 | BAC30196.1 | |||
| AK085587.1 | BAC39480.1 | |||
| BC099408.1 | AAH99408.1 | |||
| BC128494.1 | ||||
| BC128495.1 | ||||
| DV650789.1 |

Genomic Map