Assay Details
Target Gene Details
Entrez Gene ID: | 66715 |
Gene Name: | HEN1 methyltransferase homolog 1 (Arabidopsis) |
Gene Aliases: |
4921515J06Rik, Hen1, mHEN1 |
Location: |
Chr.3:108938542-108960776 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Henmt1 | NM_001078646.1 | 3 | 240 | NP_001072114.1 |
| NM_025723.2 | 3 | 555 | NP_079999.2 | |
| AK014900.1 | 3 | 523 | ||
| AK038994.1 | 3 | 241 | BAC30196.1 | |
| AK085587.1 | 3 | 555 | BAC39480.1 | |
| BC099408.1 | 2 | 221 | AAH99408.1 | |
| BC128494.1 | ||||
| BC128495.1 | ||||
| DV650789.1 | 3 | 622 |

Genomic Map