Assay Details
Target Gene Details
Entrez Gene ID: | 66715 |
Gene Name: | HEN1 methyltransferase homolog 1 (Arabidopsis) |
Gene Aliases: |
4921515J06Rik, Hen1, mHEN1 |
Location: |
Chr.3:108938542-108960776 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Henmt1 | NM_001078646.1 | 6 | 649 | NP_001072114.1 |
| NM_025723.2 | 6 | 964 | NP_079999.2 | |
| AK014900.1 | 6 | 930 | ||
| AK038994.1 | 6 | 650 | BAC30196.1 | |
| AK085587.1 | 6 | 964 | BAC39480.1 | |
| BC099408.1 | 5 | 630 | AAH99408.1 | |
| BC128494.1 | 4 | 445 | ||
| BC128495.1 | 4 | 445 |

Genomic Map