Assay Details
Target Gene Details
Entrez Gene ID: | 277343 |
Gene Name: | WAP four-disulfide core domain 8 |
Gene Aliases: |
Gm706 |
Location: |
Chr.2:164596458-164613626 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wfdc8 | NM_001080550.2 | 6 | 1023 | NP_001074019.1 |
| NM_001276431.1 | 6 | 1023 | NP_001263360.1 | |
| NM_001276432.1 | 6 | 1852 | NP_001263361.1 | |
| AK136599.1 | 6 | 1853 | ||
| AY669506.1 | 6 | 1023 | AAU89496.1 | |
| AY757956.1 | 6 | 1023 | AAU93641.1 |

Genomic Map