Assay Details
Target Gene Details
Entrez Gene ID: | 66641 |
Gene Name: | suppressor of IKBKE 1 |
Gene Aliases: |
2810005O12Rik, 5730470L24Rik, AI450236, AI839862, Sike, Sikeb |
Location: |
Chr.3:102995740-103003914 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sike1 | NM_025679.3 | NP_079955.1 | ||
| AK012679.1 | BAB28405.1 | |||
| AK014113.1 | BAB29165.1 | |||
| AK014247.1 | BAB29223.1 | |||
| AK017452.1 | ||||
| AK017694.1 | BAB30879.1 | |||
| AK019264.1 | BAB31636.1 | |||
| AK033553.1 | BAC28353.1 | |||
| AK035590.1 | 1 | 42 | BAC29116.1 | |
| AK038295.1 | ||||
| BC004090.1 | AAH04090.1 |

Genomic Map