Assay Details
Target Gene Details
Entrez Gene ID: | 21818 |
Gene Name: | transglutaminase 3, E polypeptide |
Gene Aliases: |
AI893889, TG E, TG(E), TGE, TGase E, TGase-3 |
Location: |
Chr.2:129987794-130050399 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgm3 | NM_009374.3 | 5 | 664 | NP_033400.2 |
| BC129890.1 | 5 | 630 | ||
| BC129891.1 | 5 | 630 | ||
| L10385.1 | 5 | 655 | AAA40421.1 |
More Information
Set Membership: |
|

Genomic Map