Assay Details
Target Gene Details
Entrez Gene ID: | 100502700 |
Gene Name: | expressed sequence AV039307 |
Gene Aliases: |
- |
Location: |
Chr.2:120850656-120866690 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AV039307 | NR_038349.1 | |||
| NR_038350.1 | ||||
| AK006022.1 | BAB24370.1 | |||
| AK015969.1 |
Target Gene Details
Entrez Gene ID: | 22222 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 1 |
Gene Aliases: |
AI504731 |
Location: |
Chr.2:120860272-120970729 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 45 - Exon 46 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubr1 | NM_009461.2 | NP_033487.2 | ||
| AF061555.1 | AAC40165.1 | |||
| AK078173.1 | BAC37160.1 |
More Information
Set Membership: |
|

Genomic Map