Assay Details
Target Gene Details
Entrez Gene ID: | 640543 |
Gene Name: | transglutaminase 7 |
Gene Aliases: |
TGMZ, TGz |
Location: |
Chr.2:121093498-121127516 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgm7 | NM_001160424.1 | NP_001153896.1 |
More Information
Set Membership: |
|

Genomic Map