Assay Details
Target Gene Details
Entrez Gene ID: | 50754 |
Gene Name: | F-box and WD-40 domain protein 7 |
Gene Aliases: |
1110001A17Rik, AGO, Cdc4, Fbw7, Fbwd6, Fbx30, Fbxo30, Fbxw6, SEL-10 |
Location: |
Chr.3:84815577-84979198 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxw7 | NM_001177773.1 | NP_001171244.1 | ||
| NM_001177774.1 | NP_001171245.1 | |||
| NM_080428.3 | NP_536353.2 | |||
| AB221593.1 | ||||
| AF391192.1 | AAL40928.1 | |||
| AF427101.1 | AAL50052.1 | |||
| AK032174.1 | BAC27743.1 | |||
| BC085184.1 | ||||
| BC131648.1 |
More Information
Set Membership: |
|

Genomic Map