Assay Details
Target Gene Details
Entrez Gene ID: | 14616 |
Gene Name: | gap junction protein, alpha 8 |
Gene Aliases: |
Aey5, Cnx50, Cx50, Lop10 |
Location: |
Chr.3:96913566-96926051 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gja8 | NM_008123.3 | 1 | 78 | NP_032149.1 |
| BC120513.1 | ||||
| BC125569.1 | ||||
| CV557810.1 | 1 | 79 | ||
| GU363514.1 | 1 | 47 |
More Information
Set Membership: |
|

Genomic Map