Assay Details
Target Gene Details
Entrez Gene ID: | 329735 |
Gene Name: | RIKEN cDNA 4933431E20 gene |
Gene Aliases: |
- |
Location: |
Chr.3:107888850-107896213 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933431E20Rik | NR_015459.1 | 3 | 3286 | |
| AK045063.1 | 1 | 471 | ||
| AK134641.1 | 1 | 368 | BAE22222.1 | |
| AK147824.1 | 4 | 2883 | BAE28162.1 |
Target Gene Details
Entrez Gene ID: | 99662 |
Gene Name: | EPS8-like 3 |
Gene Aliases: |
AI504489, Eps8r3 |
Location: |
Chr.3:107873860-107892900 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eps8l3 | NM_133867.2 | 15 | 1514 | NP_598628.1 |
| AK136584.1 | 15 | 1511 | ||
| AY074932.1 | 15 | 1530 | AAL76121.1 | |
| BC014734.1 | 15 | 1502 | AAH14734.1 |
More Information
Set Membership: |
|

Genomic Map