Assay Details
Target Gene Details
Entrez Gene ID: | 66371 |
Gene Name: | charged multivesicular body protein 4C |
Gene Aliases: |
2010012P02Rik, 2210015K02Rik, 2310010I16Rik, Shax3, Snf7-3 |
Location: |
Chr.3:10366973-10391005 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp4c | NM_025519.2 | NP_079795.1 | ||
| AK009273.1 | BAB26186.1 | |||
| AK010707.1 | BAB27133.1 | |||
| AK145033.1 | BAE26197.1 | |||
| AK160189.1 | BAE35682.1 | |||
| AK166371.1 | BAE38737.1 | |||
| BC115983.1 | ||||
| BC117538.1 |
More Information
Set Membership: |
|

Genomic Map