Assay Details
Target Gene Details
Entrez Gene ID: | 54195 |
Gene Name: | guanylate cyclase 1, soluble, beta 3 |
Gene Aliases: |
GC-S-beta-1, GCbeta1 |
Location: |
Chr.3:82032004-82074711 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gucy1b3 | NM_001161796.1 | NP_001155268.1 | ||
| NM_017469.4 | NP_059497.1 | |||
| AF020339.1 | AAB94876.1 | |||
| AF297083.1 | AAG17447.1 | |||
| AK139406.1 | BAE23996.1 | |||
| AK147128.1 | ||||
| BC050945.1 | AAH50945.1 | |||
| BC056995.1 | AAH56995.1 |
More Information
Set Membership: |
|

Genomic Map