Assay Details
Target Gene Details
Entrez Gene ID: | 102636083 |
Gene Name: | Mecom adjacent non-protein coding RNA |
Gene Aliases: |
Manr |
Location: |
Chr.3:29891017-29924191 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mannr | NR_110437.1 | |||
| AK147070.1 |

Genomic Map